Learn More
Sodium propionate, 99%, Thermo Scientific Chemicals
$71.26 - $188.53
Chemical Identifiers
CAS | 137-40-6 |
---|---|
Molecular Formula | C3H5NaO2 |
Molecular Weight (g/mol) | 96.061 |
MDL Number | MFCD00002759 |
InChI Key | JXKPEJDQGNYQSM-UHFFFAOYSA-M |
Synonym | sodium propionate, sodium propanoate, propionic acid sodium salt, propanoic acid, sodium salt, napropion, ocuseptine, deketon, impedex, keenate, propiofar |
PubChem CID | 2723816 |
IUPAC Name | sodium;propanoate |
SMILES | CCC(=O)[O-].[Na+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1744036
|
Thermo Scientific Chemicals
A1744036 |
500 g |
Each for $71.26
|
|
|||||
AAA174400E
|
Thermo Scientific Chemicals
A174400E |
2500 g |
Each for $188.53
|
|
|||||
Description
Sodium propionate is used as a food preservative, as a mold inhibitor in bakery products and in insect cell culture. It is also used in pharmaceuticals, cosmetics, agriculture and animal feed as preservative. It acts as a food additive. Further, it inhibits the growth of lactic acid bacteria.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium propionate is used as a food preservative, as a mold inhibitor in bakery products and in insect cell culture. It is also used in pharmaceuticals, cosmetics, agriculture and animal feed as preservative. It acts as a food additive. Further, it inhibits the growth of lactic acid bacteria.
Solubility
Soluble in water. Slightly soluble in alcohol.
Notes
Hygroscopic. Incompatible with strong oxidizing agents.
Chemical Identifiers
137-40-6 | |
96.061 | |
JXKPEJDQGNYQSM-UHFFFAOYSA-M | |
2723816 | |
CCC(=O)[O-].[Na+] |
C3H5NaO2 | |
MFCD00002759 | |
sodium propionate, sodium propanoate, propionic acid sodium salt, propanoic acid, sodium salt, napropion, ocuseptine, deketon, impedex, keenate, propiofar | |
sodium;propanoate |
Specifications
137-40-6 | |
99% | |
CH3CH2CO2Na | |
500 g | |
14,8669 | |
Soluble in water. Slightly soluble in alcohol. | |
CCC(=O)[O-].[Na+] | |
96.061 | |
96.06 | |
Sodium propionate |
285°C to 291°C | |
C3H5NaO2 | |
MFCD00002759 | |
Hygroscopic | |
sodium propionate, sodium propanoate, propionic acid sodium salt, propanoic acid, sodium salt, napropion, ocuseptine, deketon, impedex, keenate, propiofar | |
JXKPEJDQGNYQSM-UHFFFAOYSA-M | |
sodium;propanoate | |
2723816 | |
99% |
Safety and Handling
GHS H Statement
H312
Harmful in contact with skin.
GHS P Statement
P280-P312-P363-P322-P302+P352-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
IF ON SKIN: Wash with plenty of soap and water.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 205-290-4
RTECSNumber : UF7525000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only