missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Pyruvate (White Powder), Fisher BioReagents
Supplier: Fisher BioReagents FLBP356100
Description
Glass Bottle; 100g
This sodium salt is used in buffers for molecular biology and cell culture applications.Specifications
Sodium Pyruvate | |
300°C | |
5.5 to 6.5 | |
Glass Bottle | |
CH3COCO2Na | |
sodium pyruvate, pyruvic acid sodium salt, sodium 2-oxopropanoate, pyruvic acid, sodium salt, propanoic acid, 2-oxo-, sodium salt, sodium alpha-ketopropionate, pyruvate sodium, unii-pod38aif08, 2-oxopropanoic acid sodium salt, pod38aif08 | |
CC(=O)C(=O)[O-].[Na+] | |
110.044 | |
CHEBI:50144 | |
≥99% |
113-24-6 | |
White | |
≥99 % | |
C3H3NaO3 | |
100 g | |
DAEPDZWVDSPTHF-UHFFFAOYSA-M | |
sodium;2-oxopropanoate | |
23662274 | |
110.04 | |
Powder or Solid |