missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Salicylate, 99%, Thermo Scientific Chemicals

$96.08 - $270.22

Chemical Identifiers

CAS 54-21-7
Molecular Formula C7H5NaO3
Molecular Weight (g/mol) 160.104
MDL Number MFCD00002440
InChI Key ABBQHOQBGMUPJH-UHFFFAOYSA-M
Synonym sodium salicylate, sodium 2-hydroxybenzoate, salsonin, clin, enterosalicyl, enterosalil, entrosalyl, glutosalyl, kerasalicyl, magsalyl
PubChem CID 16760658
ChEBI CHEBI:9180
IUPAC Name sodium;2-hydroxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)[O-])O.[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1705630
View Documents
Thermo Scientific Chemicals
A1705630
250 g
Each for $96.08
 
AAA170560B
View Documents
Thermo Scientific Chemicals
A170560B
1000 g
Each for $270.22
 
Description

Description

Sodium salicylate is used as an analgesic and antipyretic. It also acts as a non-steroidal anti-inflammatory drug (NSAID). Further, it is used as a phosphor for the detection of vacuum ultraviolet radiation and electrons. It serves as a neurofibromatosis kappa B inhibitor and reducer of oxidative stress. It finds applications as a drug with the function of being antifebrile and anti-rheumatic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium salicylate is used as an analgesic and antipyretic. It also acts as a non-steroidal anti-inflammatory drug (NSAID). Further, it is used as a phosphor for the detection of vacuum ultraviolet radiation and electrons. It serves as a neurofibromatosis kappa B inhibitor and reducer of oxidative stress. It finds applications as a drug with the function of being antifebrile and anti-rheumatic.

Solubility
Soluble in water, 1,4-dioxane, ethanol and glycerol.

Notes
Light sensitive. Store in a cool place. Incompatible with strong oxidizing agents, strong acidsmineral acids, metallic salts and iodine.
Specifications

Chemical Identifiers

54-21-7
160.104
ABBQHOQBGMUPJH-UHFFFAOYSA-M
16760658
sodium;2-hydroxybenzoate
C7H5NaO3
MFCD00002440
sodium salicylate, sodium 2-hydroxybenzoate, salsonin, clin, enterosalicyl, enterosalil, entrosalyl, glutosalyl, kerasalicyl, magsalyl
CHEBI:9180
C1=CC=C(C(=C1)C(=O)[O-])O.[Na+]

Specifications

54-21-7
Odorless
MFCD00002440
3732792
14,8332
Soluble in water,1,4-dioxane,ethanol and glycerol.
C1=CC=C(C(=C1)C(=O)[O-])O.[Na+]
160.104
CHEBI:9180
99%
>300°C
C7H5NaO3
250 g
Light sensitive
sodium salicylate, sodium 2-hydroxybenzoate, salsonin, clin, enterosalicyl, enterosalil, entrosalyl, glutosalyl, kerasalicyl, magsalyl
ABBQHOQBGMUPJH-UHFFFAOYSA-M
sodium;2-hydroxybenzoate
16760658
160.11
Sodium salicylate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c

H302-H319

EINECSNumber : 200-198-0

RTECSNumber : VO5075000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only