missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Salicylate (Small Crystals, Free Flowing/USP), Fisher Chemical™
Supplier: Fisher Chemical FLS396500
Specifications
Sodium Salicylate | |
200°C | |
99.5 to 100.5 % | |
C7H5NaO3 | |
MFCD00002440 | |
sodium salicylate, sodium 2-hydroxybenzoate, salsonin, clin, enterosalicyl, enterosalil, entrosalyl, glutosalyl, kerasalicyl, magsalyl | |
ABBQHOQBGMUPJH-UHFFFAOYSA-M | |
sodium;2-hydroxybenzoate | |
16760658 | |
160.1 | |
USP |
54-21-7 | |
White | |
Poly Bottle | |
2-HOC6H4COONa | |
500 g | |
Meets Requirements | |
C1=CC=C(C(=C1)C(=O)[O-])O.[Na+] | |
160.104 | |
CHEBI:9180 | |
99.5 to 100.5% | |
Solid |
Chemical Identifiers
54-21-7 | |
160.104 | |
ABBQHOQBGMUPJH-UHFFFAOYSA-M | |
16760658 | |
sodium;2-hydroxybenzoate |
C7H5NaO3 | |
MFCD00002440 | |
sodium salicylate, sodium 2-hydroxybenzoate, salsonin, clin, enterosalicyl, enterosalil, entrosalyl, glutosalyl, kerasalicyl, magsalyl | |
CHEBI:9180 | |
C1=CC=C(C(=C1)C(=O)[O-])O.[Na+] |