missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium selenate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 369960250

 View more versions of this product

Catalog No. AC369960250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium selenate
White
25 g
Na2O4Se
15, 8800
Solubility in water: 19g/L (20°C)
[Na+].[Na+].[O-][Se]([O-])(=O)=O
188.95
CHEBI:77775
98%
Glass bottle
13410-01-0
Crystalline Powder
97.5% min
MFCD00003490
sodium selenate, disodium selenate, natriumseleniat, selenic acid, disodium salt, caswell no. 791, natriumseleniat german, sel-tox sso2 and ss-20, unii-5dqp25600a, selenic acid h2seo4 , disodium salt, ccris 1259
MHQOTKLEMKRJIR-UHFFFAOYSA-L
disodium;selenate
25960
188.94
confirmed

Chemical Identifiers

13410-01-0
188.95
MHQOTKLEMKRJIR-UHFFFAOYSA-L
25960
disodium;selenate
Na2O4Se
MFCD00003490
sodium selenate, disodium selenate, natriumseleniat, selenic acid, disodium salt, caswell no. 791, natriumseleniat german, sel-tox sso2 and ss-20, unii-5dqp25600a, selenic acid h2seo4 , disodium salt, ccris 1259
CHEBI:77775
[Na+].[Na+].[O-][Se]([O-])(=O)=O
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Fatal if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED:

GHS Signal Word: Danger

EINECSNumber : 236-501-8

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only