Learn More
Sodium selenate, anhydrous, 99.8+% (metals basis)
For Use As a Reagent | CAS: 13410-01-0 | Na2O4Se | 188.95 g/mol
$66.73 - $1,034.47
Chemical Identifiers
| CAS | 13410-01-0 |
|---|---|
| Molecular Formula | Na2O4Se |
| Molecular Weight (g/mol) | 188.95 |
| MDL Number | MFCD00003490 |
| InChI Key | MHQOTKLEMKRJIR-UHFFFAOYSA-L |
| Synonym | sodium selenate, disodium selenate, natriumseleniat, selenic acid, disodium salt, caswell no. 791, natriumseleniat german, sel-tox sso2 and ss-20, unii-5dqp25600a, selenic acid h2seo4 , disodium salt, ccris 1259 |
| PubChem CID | 25960 |
| ChEBI | CHEBI:77775 |
| IUPAC Name | disodium;selenate |
| SMILES | [Na+].[Na+].[O-][Se]([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA1261306
|
Thermo Scientific Chemicals
01261306 |
5 g |
N/A
|
|
|||||
|
AA1261318
|
Thermo Scientific Chemicals
01261318 |
50 g |
N/A
|
|
|||||
|
AA1261330
|
Thermo Scientific Chemicals
01261330 |
250 g |
N/A
|
|
|||||
|
AA12613A1
|
Thermo Scientific Chemicals
012613A1 |
1 kg |
N/A
|
|
|||||
Description
Sodium selenate is used in glass industry, and produces red hue in glass. It acts as a decoloring agent in glass production. Combined with ferrous oxides, it produces colorless glass. Appropriate preparation of sodium selenate is used in multivitamins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 13410-01-0 | |
| 188.95 | |
| MHQOTKLEMKRJIR-UHFFFAOYSA-L | |
| 25960 | |
| disodium;selenate |
| Na2O4Se | |
| MFCD00003490 | |
| sodium selenate, disodium selenate, natriumseleniat, selenic acid, disodium salt, caswell no. 791, natriumseleniat german, sel-tox sso2 and ss-20, unii-5dqp25600a, selenic acid h2seo4 , disodium salt, ccris 1259 | |
| CHEBI:77775 | |
| [Na+].[Na+].[O-][Se]([O-])(=O)=O |
Specifications
| 32° (transition) | |
| Purple | |
| 5 g | |
| Na2O4Se | |
| sodium selenate, disodium selenate, natriumseleniat, selenic acid, disodium salt, caswell no. 791, natriumseleniat german, sel-tox sso2 and ss-20, unii-5dqp25600a, selenic acid h2seo4 , disodium salt, ccris 1259 | |
| MHQOTKLEMKRJIR-UHFFFAOYSA-L | |
| disodium;selenate | |
| 25960 | |
| 188.94 | |
| Hygroscopic | |
| 3.2 g/cm3 at 20°C |
| 13410-01-0 | |
| Powder | |
| 99.8+% (metals basis) | |
| MFCD00003490 | |
| Soluble in water. | |
| [Na+].[Na+].[O-][Se]([O-])(=O)=O | |
| 188.95 | |
| CHEBI:77775 | |
| Odorless | |
| White powder | |
| Sodium selenate, anhydrous |
Safety and Handling
P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c
H300-H331-H373
DOTInformation : DOT Class: 6.1, Packaging Group: I
EINECSNumber : 236-501-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only