Learn More
Sodium selenate decahydrate, 99.9% (metals basis), Thermo Scientific Chemicals
$181.36 - $527.96
Chemical Identifiers
CAS | 10102-23-5 |
---|---|
Molecular Formula | H20Na2O14Se |
Molecular Weight (g/mol) | 369.10 |
MDL Number | MFCD00149174 |
InChI Key | TYVNFQRMDPFYPC-UHFFFAOYSA-L |
Synonym | sodium selenate decahydrate, unii-0cjn4029eb, disodium decahydrate selenate, disodium selenate decahydrate, acmc-1c452, sodium selenate decahydrate, technical grade, selenic acid, disodiumsalt, decahydrate 9ci, sodium selenate decahydrate trace metals basis |
PubChem CID | 202320 |
IUPAC Name | disodium;selenate;decahydrate |
SMILES | O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-][Se]([O-])(=O)=O |
Description
Sodium selenate decahydrate is used as a feed additive, as a mineral supplement, as analytical reagents and in horticultural applications. Further, it is used in manufacturing of glass to counter the color caused by iron impurities. It is also employed in the prevention of abnormal tissue growth. In addition to this, it is used as a micronutrient which prevents a deficiency of copper, manganese and selenium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10102-23-5 | |
369.10 | |
TYVNFQRMDPFYPC-UHFFFAOYSA-L | |
202320 | |
O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-][Se]([O-])(=O)=O |
H20Na2O14Se | |
MFCD00149174 | |
sodium selenate decahydrate, unii-0cjn4029eb, disodium decahydrate selenate, disodium selenate decahydrate, acmc-1c452, sodium selenate decahydrate, technical grade, selenic acid, disodiumsalt, decahydrate 9ci, sodium selenate decahydrate trace metals basis | |
disodium;selenate;decahydrate |
Specifications
35°C (decomposition) | |
Crystalline | |
99.9% (metals basis) | |
MFCD00149174 | |
14,8671 | |
Soluble in water. | |
O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-][Se]([O-])(=O)=O | |
369.10 | |
369.09 (188.94 Anhydrous) | |
1.6 g/mL | |
(metals basis) |
10102-23-5 | |
25 g | |
H20Na2O14Se | |
UN2630 | |
sodium selenate decahydrate, unii-0cjn4029eb, disodium decahydrate selenate, disodium selenate decahydrate, acmc-1c452, sodium selenate decahydrate, technical grade, selenic acid, disodiumsalt, decahydrate 9ci, sodium selenate decahydrate trace metals basis | |
TYVNFQRMDPFYPC-UHFFFAOYSA-L | |
disodium;selenate;decahydrate | |
202320 | |
99.9% | |
Sodium selenate decahydrate |
Safety and Handling
GHS H Statement
H301-H331-H373
P260-P264b-P270-P271-P301+P310-P304+P340-P311-P314-P330-P501c
H301+H331-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: SELENATES
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only