missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium sticks, coated in film of protective hydrocarbon oil, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1328536

 View more versions of this product

Catalog No. AAL1328536


Explore available promotions

Description

Description

Sodium sticks, in mineral oil is used for reviews of Na metal - ammonia (Birch) reductions, for preparation of sodamide in liquid ammonia and for use in an acyloin synthesis promoted by sonication.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium sticks, in mineral oil is used for reviews of Na metal - ammonia (Birch) reductions, for preparation of sodamide in liquid ammonia and for use in an acyloin synthesis promoted by sonication.

Solubility
Reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Handle and store under inert gas. It is sensitive to air.
Specifications

Specifications

Sodium sticks
98°C
881°C
MFCD00085307
500 g
14,8570
Reacts with water.
[Na]
22.99
CHEBI:26708
Air and moisture sensitive
in mineral oil
7440-23-5
Na
UN1428
>110°C (230°F)
natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot
KEAYESYHFKHZAL-UHFFFAOYSA-N
sodium
5360545
99%

Chemical Identifiers

7440-23-5
22.99
KEAYESYHFKHZAL-UHFFFAOYSA-N
5360545
sodium
Na
MFCD00085307
natrium, sodium-23, sodio, metal, sodio spanish, liquid alloy, unii-9nez333n27, hsdb 687, unii-23j3bhr95o, ingot
CHEBI:26708
[Na]
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H260-H314
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.

P223-P231+P232-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P335+P334-P363-P370+P378q-P501c

H260-H304-H314-H335

DOTInformation : Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: SODIUM

EINECSNumber : 231-132-9

RTECSNumber : VY0686000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only