missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Sulfate Anhydrous, Certified ACS, Suitable for Pesticide Residue Analysis (Granular, 10-60 mesh), Fisher Chemical™

$276.17 - $12807.58

Chemical Identifiers

CAS 7757-82-6
Molecular Formula Na2O4S
Molecular Weight (g/mol) 142.04
MDL Number MFCD00003504
InChI Key PMZURENOXWZQFD-UHFFFAOYSA-L
Synonym sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
PubChem CID 24436
ChEBI CHEBI:32149
IUPAC Name disodium sulfate
SMILES [Na+].[Na+].[O-]S([O-])(=O)=O
View More Specs

Products 6
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
S415-500
SDS View Documents
Fisher Chemical
S415500
500 g Glass Bottle
Each for $276.17
Case of 6 Each for $1,657.02
 
S415-200LB
SDS Accessories View Documents
Thermo Fisher Scientific
S415200LB
200 lb. Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
S415-1
SDS View Documents
Thermo Fisher Scientific
S4151
1 kg Poly Bottle
Each for $403.00
Case of 6 Each for $2,418.00
 
S415-212
SDS View Documents
Thermo Fisher Scientific
S415212
2.5 kg Glass Bottle
Each for $768.75
Case of 4 Each for $3,075.00
 
S415-10S Edge
SDS View Documents
Thermo Fisher Scientific
S41510
10 kg Poly Pail
Each for $1,880.83
Edge
 
S41550
SDS View Documents
Fisher Chemical
S41550
50 kg Fiber Drum
Each for $5,366.84
 
Description

Description

Specifications

Chemical Identifiers

7757-82-6
142.04
PMZURENOXWZQFD-UHFFFAOYSA-L
24436
disodium sulfate
Na2O4S
MFCD00003504
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
CHEBI:32149
[Na+].[Na+].[O-]S([O-])(=O)=O

Specifications

884°C
White
Solid
≥99.0 %
MFCD00003504
PMZURENOXWZQFD-UHFFFAOYSA-L
disodium sulfate
24436
142.04
Suitable for pesticide residue analysis
0.5% max.
Decomposition Information: >890°C
7757-82-6
5 to 8
500 g
Na2O4S
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
[Na+].[Na+].[O-]S([O-])(=O)=O
142.04
CHEBI:32149
≥99.0%
Pass Test
Glass Bottle
Sodium Sulfate Anhydrous
Videos
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause skin, eye, and respiratory tract irritation. The toxicological properties have not been fully investigated. Hygroscopic.
NFPA
Health: 1
Flammability: 1
Instability: 0

SIGNAL WORD: CAUTION!

SDS
missing translation for 'documents'

missing translation for 'documents'