missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Sulfate, Anhydrous, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
Supplier: Avantor J.T.Baker 389801
Specifications
Sodium Sulfate | |
7757-82-6 | |
500 g | |
MFCD00003504 | |
PMZURENOXWZQFD-UHFFFAOYSA-L | |
disodium;sulfate | |
24436 | |
142.04 | |
Wide Mouth Amber Glass Bottle |
Anhydrous | |
Powder | |
Na2O4S | |
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1 | |
[Na+].[Na+].[O-]S([O-])(=O)=O | |
142.04 | |
CHEBI:32149 | |
BAKER ANALYZED™ ACS Reagent | |
1L = 2.68kg |
Chemical Identifiers
7757-82-6 | |
142.04 | |
PMZURENOXWZQFD-UHFFFAOYSA-L | |
24436 | |
disodium;sulfate |
Na2O4S | |
MFCD00003504 | |
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1 | |
CHEBI:32149 | |
[Na+].[Na+].[O-]S([O-])(=O)=O |