missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Sulfate, Anhydrous, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

High quality chemicals for laboratory and specialized industrial use

Supplier:  Avantor J.T.Baker 389805

 View more versions of this product

Catalog No. 02004241


Explore available promotions

Description

Description

Specifications

Specifications

Sodium Sulfate
7757-82-6
2.5 kg
MFCD00003504
PMZURENOXWZQFD-UHFFFAOYSA-L
disodium;sulfate
24436
142.04
Wide Mouth Amber Glass Bottle
Anhydrous
Powder
Na2O4S
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
[Na+].[Na+].[O-]S([O-])(=O)=O
142.04
CHEBI:32149
BAKER ANALYZED™ ACS Reagent
1L = 2.68kg

Chemical Identifiers

7757-82-6
142.04
PMZURENOXWZQFD-UHFFFAOYSA-L
24436
disodium;sulfate
Na2O4S
MFCD00003504
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
CHEBI:32149
[Na+].[Na+].[O-]S([O-])(=O)=O
Videos
SDS
Documents

Documents

Promotions

Promotions