missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Sulfate Anhydrous (Powder/USP/FCC), Fisher Chemical™

Supplier:  Thermo Fisher Scientific FLS429500

 View more versions of this product

Catalog No. S429-500

Explore available promotions

Description

Description

Specifications

Specifications

Sodium Sulfate Anhydrous
7757-82-6
5 to 8
500 g
MFCD00003504
PMZURENOXWZQFD-UHFFFAOYSA-L
disodium sulfate
24436
142.04
FCC/USP
0.5% max. (USP), 0.5% max. (USP), 1% max. (FCC), 1% max. (FCC)
Decomposition Information: >890°C
884°C
White
Solid
Na2O4S
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
[Na+].[Na+].[O-]S([O-])(=O)=O
142.04
CHEBI:32149
99.0 to 100.5% (FCC)
Pass Test (FCC), Pass Test (USP), Pass Test (USP/FCC)
Poly Bottle

Chemical Identifiers

7757-82-6
142.04
PMZURENOXWZQFD-UHFFFAOYSA-L
24436
disodium sulfate
Na2O4S
MFCD00003504
sodium sulfate, disodium sulfate, salt cake, thenardite, sodium sulfate anhydrous, sodium sulphate, sodium sulfate, anhydrous, sulfuric acid disodium salt, disodium sulphate, sodium sulfate 2:1
CHEBI:32149
[Na+].[Na+].[O-]S([O-])(=O)=O
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause skin, eye, and respiratory tract irritation. The toxicological properties have not been fully investigated. Hygroscopic.
NFPA
Health: 1
Flammability: 1
Instability: 0

SIGNAL WORD: CAUTION!

SDS Documents

Documents

Promotions

Promotions