missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium tetraborate decahydrate, 99.5%, for analysis, Thermo Scientific Chemicals
$101.02 - $291.49
Chemical Identifiers
CAS | 1303-96-4 |
---|---|
Molecular Formula | B4Na2O7·10H2O |
Molecular Weight (g/mol) | 381.36 |
MDL Number | MFCD00149193 |
Synonym | Borax, Sodium borate decahydrate |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC205950010
|
Thermo Scientific Chemicals
205950010 |
1 kg | Plastic Bottle |
Each for $101.02
|
|
||||
AC205950050
|
Thermo Scientific Chemicals
205950050 |
5 kg | Plastic drum |
Each for $291.49
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Biological buffer for analysisChemical Identifiers
1303-96-4 | |
381.36 | |
Borax, Sodium borate decahydrate |
B4Na2O7·10H2O | |
MFCD00149193 |
Specifications
75.0°C | |
1303-96-4 | |
Fine Crystalline Powder | |
99.5% | |
Na2B4O7·10H2O | |
04,461 | |
Borax, Sodium borate decahydrate | |
381.36 | |
99.5% | |
Plastic Bottle |
White | |
9 to 9.5 (5% soln. at 20°C) | |
1 kg | |
B4Na2O7·10H2O | |
MFCD00149193 | |
15,8726 | |
Solubility in water: 60g/L (20°C). Other solubilities: soluble in glycerol, insoluble in alcohol | |
381.36 | |
Analytical | |
Sodium tetraborate decahydrate |
Safety and Handling
GHS H Statement
May damage fertility.
May damage the unborn child.
GHS P Statement
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
GHS Signal Word: Danger
RUO – Research Use Only