missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium tetrafluoroborate, 97+%, extra pure, Thermo Scientific Chemicals

$92.37 - $134.21

Chemical Identifiers

CAS 13755-29-8
Molecular Formula BF4Na
Molecular Weight (g/mol) 109.79
MDL Number MFCD00003515
InChI Key KGJZTOFHXCFQIV-UHFFFAOYSA-N
Synonym sodium tetrafluoroborate, sodium fluoborate, sodium fluoroborate, sodium borofluoride, sodium boron fluoride, apreton r, sodium boron tetrafluoride, sodium fluoroborate nabf4, sodium tetrafluoroborate 1-, sodium tetrafluroborate
PubChem CID 4343483
SMILES [Na+].F[B-](F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC192120250
SDS View Documents
Thermo Scientific Chemicals
192120250
25 g Plastic bottle
Each for $92.37
 
AC192125000
SDS View Documents
Thermo Scientific Chemicals
192125000
500 g Plastic bottle
Each for $134.21
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
Specifications

Chemical Identifiers

13755-29-8
109.79
KGJZTOFHXCFQIV-UHFFFAOYSA-N
4343483
BF4Na
MFCD00003515
sodium tetrafluoroborate, sodium fluoborate, sodium fluoroborate, sodium borofluoride, sodium boron fluoride, apreton r, sodium boron tetrafluoride, sodium fluoroborate nabf4, sodium tetrafluoroborate 1-, sodium tetrafluroborate
[Na+].F[B-](F)(F)F

Specifications

384°C
Brown to White
25 g
BF4Na
MFCD00003515
sodium tetrafluoroborate, sodium fluoborate, sodium fluoroborate, sodium borofluoride, sodium boron fluoride, apreton r, sodium boron tetrafluoride, sodium fluoroborate nabf4, sodium tetrafluoroborate 1-, sodium tetrafluroborate
KGJZTOFHXCFQIV-UHFFFAOYSA-N
sodium tetrafluoroboranuide
4343483
≥97%
Plastic bottle
Sodium tetrafluoroborate, 98%
13755-29-8
Powder, Needles or Crystals
97+%
NaBF4
15, 8753
Solubility in water: 973g/L (20°C). Other solubilities: sparingly soluble in alcohol
[Na+].F[B-](F)(F)F
109.79
109.79
Extra Pure
2.47
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 237-340-6

RTECSNumber : ED2975000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only