missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate, 97%, may cont. 1-5% water, Thermo Scientific Chemicals

Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate, 97%, C32H12BF24Na, CAS Number-79060-88-1

$312.74 - $3138.93

Chemical Identifiers

CAS 79060-88-1
Molecular Formula C32H12BF24Na
Molecular Weight (g/mol) 886.209
MDL Number MFCD00043323
InChI Key LTGMONZOZHXAHO-UHFFFAOYSA-N
Synonym sodium tetrakis 3,5-bis trifluoromethyl phenyl borate, nabarf, sodium tetrakis 3,5-bis trifluoromethyl phenyl boranuide, sodium2-sulfonatoethylmethacrylate, sodium tetrakis 3,5-bis trifluoromethyl phenyl bor, sodium tetrakis 3,5-bis-trifluoromethylphenyl borate, sodiotetrakis 3,5-bis trifluoromethyl phenyl boron v, sodium tetrakis 3,5-bis trifluoromethyl phenyl-borate, sodium tetrakis 3,5-bis trifluoromethyl phenyl borate 1-
PubChem CID 23681909
IUPAC Name sodium;tetrakis[3,5-bis(trifluoromethyl)phenyl]boranuide
SMILES [B-](C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)(C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F)(C3=CC(=CC(=C3)C(F)(F)F)C(F)(F)F)C4=CC(=CC(=C4)C(F)(F)F)C(F)(F)F.[Na+]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH3001403
View Documents
Thermo Scientific Chemicals
H3001403
1 g
Each for $312.74
 
AAH3001406
View Documents
Thermo Scientific Chemicals
H3001406
5 g
Each for $997.07
 
AAH3001414
View Documents
Thermo Scientific Chemicals
H3001414
25 g
Each for $3,138.93
 
Description

Description

Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate is a lipophilic additive of tetraphenylborate and it is used for solvent extraction of cations and solvent polymeric membrane electrodes and optodes. It is also used as an anionic phase-transfer catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate is a lipophilic additive of tetraphenylborate and it is used for solvent extraction of cations and solvent polymeric membrane electrodes and optodes. It is also used as an anionic phase-transfer catalyst.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

79060-88-1
886.209
LTGMONZOZHXAHO-UHFFFAOYSA-N
23681909
[B-](C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)(C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F)(C3=CC(=CC(=C3)C(F)(F)F)C(F)(F)F)C4=CC(=CC(=C4)C(F)(F)F)C(F)(F)F.[Na+]
C32H12BF24Na
MFCD00043323
sodium tetrakis 3,5-bis trifluoromethyl phenyl borate, nabarf, sodium tetrakis 3,5-bis trifluoromethyl phenyl boranuide, sodium2-sulfonatoethylmethacrylate, sodium tetrakis 3,5-bis trifluoromethyl phenyl bor, sodium tetrakis 3,5-bis-trifluoromethylphenyl borate, sodiotetrakis 3,5-bis trifluoromethyl phenyl boron v, sodium tetrakis 3,5-bis trifluoromethyl phenyl-borate, sodium tetrakis 3,5-bis trifluoromethyl phenyl borate 1-
sodium;tetrakis[3,5-bis(trifluoromethyl)phenyl]boranuide

Specifications

79060-88-1
C32H12BF24Na
1 g
Soluble in water.
[B-](C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)(C2=CC(=CC(=C2)C(F)(F)F)C(F)(F)F)(C3=CC(=CC(=C3)C(F)(F)F)C(F)(F)F)C4=CC(=CC(=C4)C(F)(F)F)C(F)(F)F.[Na+]
886.209
886.2
Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate
310°C (decomposition)
MFCD00043323
sodium tetrakis 3,5-bis trifluoromethyl phenyl borate, nabarf, sodium tetrakis 3,5-bis trifluoromethyl phenyl boranuide, sodium2-sulfonatoethylmethacrylate, sodium tetrakis 3,5-bis trifluoromethyl phenyl bor, sodium tetrakis 3,5-bis-trifluoromethylphenyl borate, sodiotetrakis 3,5-bis trifluoromethyl phenyl boron v, sodium tetrakis 3,5-bis trifluoromethyl phenyl-borate, sodium tetrakis 3,5-bis trifluoromethyl phenyl borate 1-
LTGMONZOZHXAHO-UHFFFAOYSA-N
sodium;tetrakis[3,5-bis(trifluoromethyl)phenyl]boranuide
23681909
97%
may contain 1 to 5% water
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only