missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium tetrakis(4-fluorophenyl)borate dihydrate, 98%, Thermo Scientific Chemicals

$277.58 - $2658.92

Chemical Identifiers

CAS 207683-22-5
Molecular Formula C24H20BF4NaO2
Molecular Weight (g/mol) 450.215
MDL Number MFCD00149598
InChI Key MSDGDEJOIBMWJD-UHFFFAOYSA-N
Synonym sodium tetrakis 4-fluorophenyl borate dihydrate, cesibor, tetrakis 4-fluorophenyl boron sodium, sodium tetrakis 4-fluorophenyl boranuide dihydrate, sodium tetrakis 4-fluorophenyl borate dihydrate nt, sodium tetrakis 4-fluorophenyl borate dihydrate, selectophore tm
PubChem CID 45073643
IUPAC Name sodium;tetrakis(4-fluorophenyl)boranuide;dihydrate
SMILES [B-](C1=CC=C(C=C1)F)(C2=CC=C(C=C2)F)(C3=CC=C(C=C3)F)C4=CC=C(C=C4)F.O.O.[Na+]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH55952MD
View Documents
Thermo Scientific Chemicals
H55952MD
250 mg
Each for $277.58
 
AAH5595203
View Documents
Thermo Scientific Chemicals
H5595203
1 g
Each for $774.30
 
AAH5595206
View Documents
Thermo Scientific Chemicals
H5595206
5 g
Each for $2,658.92
 
Description

Description

Sodium tetrakis(4-fluorophenyl)borate dihydrate, is used as a reagent for the two-phase titration of nonionic surfactants in the presence of anionic surfactants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium tetrakis(4-fluorophenyl)borate dihydrate, is used as a reagent for the two-phase titration of nonionic surfactants in the presence of anionic surfactants.

Notes
Hygroscopic. Incompatible withstrong oxidizing agents.
Specifications

Chemical Identifiers

207683-22-5
450.215
MSDGDEJOIBMWJD-UHFFFAOYSA-N
45073643
[B-](C1=CC=C(C=C1)F)(C2=CC=C(C=C2)F)(C3=CC=C(C=C3)F)C4=CC=C(C=C4)F.O.O.[Na+]
C24H20BF4NaO2
MFCD00149598
sodium tetrakis 4-fluorophenyl borate dihydrate, cesibor, tetrakis 4-fluorophenyl boron sodium, sodium tetrakis 4-fluorophenyl boranuide dihydrate, sodium tetrakis 4-fluorophenyl borate dihydrate nt, sodium tetrakis 4-fluorophenyl borate dihydrate, selectophore tm
sodium;tetrakis(4-fluorophenyl)boranuide;dihydrate

Specifications

207683-22-5
MFCD00149598
4285808
MSDGDEJOIBMWJD-UHFFFAOYSA-N
sodium;tetrakis(4-fluorophenyl)boranuide;dihydrate
45073643
98%
C24H20BF4NaO2
250 mg
sodium tetrakis 4-fluorophenyl borate dihydrate, cesibor, tetrakis 4-fluorophenyl boron sodium, sodium tetrakis 4-fluorophenyl boranuide dihydrate, sodium tetrakis 4-fluorophenyl borate dihydrate nt, sodium tetrakis 4-fluorophenyl borate dihydrate, selectophore tm
[B-](C1=CC=C(C=C1)F)(C2=CC=C(C=C2)F)(C3=CC=C(C=C3)F)C4=CC=C(C=C4)F.O.O.[Na+]
450.215
450.21 (414.18 Anhydrous)
Sodium tetrakis(4-fluorophenyl)borate dihydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only