missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium thiocyanate, ACS, 98.0% min, Thermo Scientific Chemicals

$90.03 - $580.73

Chemical Identifiers

CAS 540-72-7
Molecular Formula CNNaS
Molecular Weight (g/mol) 81.07
MDL Number MFCD00011123
InChI Key VGTPCRGMBIAPIM-UHFFFAOYSA-M
Synonym sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased
PubChem CID 516871
ChEBI CHEBI:30952
SMILES [Na+].[S-]C#N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3338822
View Documents
Thermo Scientific Chemicals
03338822
100 g
Each for $90.03
 
AA3338836
View Documents
Thermo Scientific Chemicals
03338836
500 g
Each for $199.05
 
AA33388A3
View Documents
Thermo Scientific Chemicals
033388A3
2 kg
Each for $580.73
 
Description

Description

Sodium thiocyanate is used as a precursor for the synthesis of pharmaceuticals and specialty chemicals. It is used to prepare alkylthiocyanates by reacting with alkyl halides. It is involved nucleophilic substitution reactions with phase-transfer catalysts. It is a common laboratory reagent used to test presence of Fe3+ ions. It acts as a spinning solvent for polyacrylonitrile in fiber industry, photographic stabilizer, corrosion inhibitor, polymer stabilizer and hardening accelerator for cement and concrete mixes. Further, it is also used in both the crystallization and purification of proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

540-72-7
81.07
VGTPCRGMBIAPIM-UHFFFAOYSA-M
516871
[Na+].[S-]C#N
CNNaS
MFCD00011123
sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased
CHEBI:30952

Specifications

287°C
White
100 g
CNNaS
3594965
Soluble in water,acetone,alcohol.Soluble in water,acetone,alcohols and ammonia.
[Na+].[S-]C#N
81.07
CHEBI:30952
ACS
White crystalline/chunks
Sodium thiocyanate, For analysis ACS
540-72-7
Crystals and/or Chunks
98% min.
MFCD00011123
sodium thiocyanate, sodium rhodanate, sodium sulfocyanate, sodium rhodanide, thiocyanic acid, sodium salt, thiocyanate sodium, scyan, sodium isothiocyanate, nascn, haimased
VGTPCRGMBIAPIM-UHFFFAOYSA-M
sodium;thiocyanate
516871
81.07
Hygroscopic; Light sensitive
1.74 g/cm3 at 20°C
Videos
Safety and Handling

Safety and Handling

P260-P264b-P270-P271-P280-P302+P352-P304+P340-P314-P330-P362-P501c

H302+H312+H332-H373

EINECSNumber : 208-754-4

RTECSNumber : XL2275000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only