Learn More
Sodium thiomethoxide, 90+%, technical, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 469890250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium thiomethoxide | |
90+% | |
CH3SNa | |
Solubility aq. soln.: freely soluble. | |
[Na+].C[S-] | |
70.08 | |
Technical |
5188-07-8 | |
CH3NaS | |
25 g | |
RMBAVIFYHOYIFM-UHFFFAOYSA-M | |
sodium methylsulfanide | |
70.09 |
Chemical Identifiers
5188-07-8 | |
70.08 | |
sodium methylsulfanide |
CH3NaS | |
RMBAVIFYHOYIFM-UHFFFAOYSA-M | |
[Na+].C[S-] |
Safety and Handling
GHS H Statement Flammable solid.
Toxic if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement Keep away from heat, hot surfaces, sparks, open flames and other ignition sources.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water or shower
GHS Signal Word: Danger
EINECSNumber : 225-969-9
RUO – Research Use Only