missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium thiosulfate, 0.1 N standard solution, Thermo Scientific Chemicals
$79.02 - $1046.98
Specifications
Chemical Name or Material | Sodium thiosulfate |
---|---|
CAS | 7772-98-7,7732-18-5 |
Color | Colorless |
Physical Form | Liquid |
Molecular Formula | Na2O3S2 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC124270010
|
Thermo Scientific Chemicals
124270010 |
1 L | Plastic bottle |
Each for $79.02
|
|
||||
AC124270025
|
Thermo Scientific Chemicals
124270025 |
2.5 L | Plastic Bottle |
Each for $164.20
|
|
||||
AC124270250
|
Thermo Scientific Chemicals
124270250 |
25 L | Plastic Drum |
Each for $1,046.98
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent, Volumetric standard solutionSpecifications
Sodium thiosulfate | |
Colorless | |
Na2O3S2 | |
MFCD00003499 | |
sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt | |
[Na+].[Na+].[O-]S([S-])(=O)=O | |
24477 | |
0.0950 to 0.1050N (20°C) |
7772-98-7,7732-18-5 | |
Liquid | |
Na2S2O3 | |
15, 8821 | |
AKHNMLFCWUSKQB-UHFFFAOYSA-L | |
158.10 | |
158.11 |
RUO – Research Use Only