missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium thiosulfate, 98.5%, extra pure, anhydrous
CAS: 7772-98-7 | Na2O3S2 | 158.10 g/mol
$166.74 - $428.26
Identifiants chimiques
| CAS | 7772-98-7 |
|---|---|
| Molecular Formula | Na2O3S2 |
| Molecular Weight (g/mol) | 158.10 |
| MDL Number | MFCD00003499 |
| InChI Key | AKHNMLFCWUSKQB-UHFFFAOYSA-L |
| Synonym | sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt |
| PubChem CID | 24477 |
| SMILES | [Na+].[Na+].[O-]S([S-])(=O)=O |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
|
AC202875000
|
Thermo Scientific Chemicals
202875000 |
500 g | Plastic bottle |
chaque for $166.74
caisse de 6 chaque for $0.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
|
AC202870010
|
Thermo Scientific Chemicals
202870010 |
1 kg | Plastic bottle |
chaque for $235.36
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
|
AC202870025
|
Thermo Scientific Chemicals
202870025 |
2.5 kg | Plastic bottle |
chaque for $428.26
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
|||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentIdentifiants chimiques
| 7772-98-7 | |
| 158.10 | |
| AKHNMLFCWUSKQB-UHFFFAOYSA-L | |
| 24477 |
| Na2O3S2 | |
| MFCD00003499 | |
| sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt | |
| [Na+].[Na+].[O-]S([S-])(=O)=O |
Spécifications
| 48.0°C | |
| White | |
| Crystalline Powder | |
| 98% min. (on dry substance) (Iodimetry) | |
| Na2S2O3 | |
| 15,8821 | |
| Solubility in water: 700g/L (20°C). Other solubilities: practically insoluble in alcohol | |
| [Na+].[Na+].[O-]S([S-])(=O)=O | |
| 158.10 | |
| 158.1 | |
| Extra Pure | |
| Plastic bottle |
| 7772-98-7 | |
| 6.0 to 8.5 (5% soln. at 20°C) | |
| 500 g | |
| Na2O3S2 | |
| MFCD00003499 | |
| sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt | |
| AKHNMLFCWUSKQB-UHFFFAOYSA-L | |
| disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane | |
| 24477 | |
| 98.5% | |
| 2% (105°C, 2 hrs) max. | |
| Sodium thiosulfate |
Sécurité et manipulation
missing translation for 'einecsNumber' : 231-867-5
RUO – Research Use Only