missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Thiosulfate Anhydrous (Certified), Fisher Chemical™
Supplier: Fisher Chemical S446500
Specifications
| Sodium Thiosulfate Anhydrous | |
| White | |
| 6.0 to 8.5 | |
| 500 g | |
| Na2O3S2 | |
| sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt | |
| [Na+].[Na+].[O-]S([S-])(=O)=O | |
| 158.10 | |
| 158.11 | |
| Certified | |
| Poly Bottle |
| 48°C | |
| 7772-98-7 | |
| Solid | |
| ≥98 % | |
| MFCD00003499 | |
| AKHNMLFCWUSKQB-UHFFFAOYSA-L | |
| disodium sulfanidesulfonate | |
| 24477 | |
| ≥98% | |
| Pass Test | |
| Negligible |
Chemical Identifiers
| 7772-98-7 | |
| 158.10 | |
| AKHNMLFCWUSKQB-UHFFFAOYSA-L | |
| 24477 | |
| [Na+].[Na+].[O-]S([S-])(=O)=O |
| Na2O3S2 | |
| MFCD00003499 | |
| sodium thiosulfate, sodium thiosulphate, disodium thiosulfate, sodium thiosulfate anhydrous, hypo, sodiumthiosulfate, chlorine cure, chlorine control, declor-it, thiosulfuric acid, disodium salt | |
| disodium sulfanidesulfonate |
Safety and Handling