Learn More
Sodium triacetoxyborohydride, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2206014
Description
Sodium triacetoxyborohydride is used as a reagent in reductive amination of ketones, aldehydes and lactamization of carbonyl compounds with amines. It also serves as a reducing agent in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium triacetoxyborohydride is used as a reagent in reductive amination of ketones, aldehydes and lactamization of carbonyl compounds with amines. It also serves as a reducing agent in organic synthesis.
Solubility
Soluble in dimethyl sulfoxide, methanol, benzene, toluene, terahydrofuran, dioxane and methylene chloride.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, water, alcohols and acids.
Specifications
Sodium triacetoxyborohydride | |
∼110°C (decomposition) | |
MFCD00012211 | |
UN3396 | |
Moisture sensitive | |
Soluble in dimethyl sulfoxide,methanol,benzene,toluene,terahydrofuran,dioxane and methylene chloride. | |
[Na+].CC(=O)O[BH-](OC(C)=O)OC(C)=O | |
211.94 | |
95% |
56553-60-7 | |
C6H10BNaO6 | |
25 g | |
4047608 | |
c6h10bnao6, sodium triacetoxyboron 1-, pubchem12871, sodium triacetoxy borohyride, sodium triacetyloxyboron 1-, tris acetoxy hydroborate sodium salt, sodium bis acetyloxy-$l^ 2-boranuidyl acetate | |
HHYFEYBWNZJVFQ-UHFFFAOYSA-N | |
sodium bis(acetyloxy)boranuidyl acetate | |
211.94 |
Chemical Identifiers
56553-60-7 | |
211.94 | |
HHYFEYBWNZJVFQ-UHFFFAOYSA-N | |
sodium bis(acetyloxy)boranuidyl acetate |
C6H10BNaO6 | |
MFCD00012211 | |
c6h10bnao6, sodium triacetoxyboron 1-, pubchem12871, sodium triacetoxy borohyride, sodium triacetyloxyboron 1-, tris acetoxy hydroborate sodium salt, sodium bis acetyloxy-$l^ 2-boranuidyl acetate | |
[Na+].CC(=O)O[BH-](OC(C)=O)OC(C)=O |
Safety and Handling
GHS H Statement
H261-H228-H318-H335-H315
In contact with water releases flammable gas.
Flammable solid.
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
P223-P231+P232-P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P335+P334-P362-P370+P378q-P501c
H260-H315-H318
DOTInformation : Hazard Class: 4.3; Packaging Group: II
EINECSNumber : 000-000-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only