Learn More
Sodium trichloroacetate, 97%, Thermo Scientific Chemicals
$70.85 - $672.03
Chemical Identifiers
CAS | 650-51-1 |
---|---|
Molecular Formula | C2Cl3NaO2 |
Molecular Weight (g/mol) | 185.36 |
MDL Number | MFCD00064198 |
InChI Key | SAQSTQBVENFSKT-UHFFFAOYSA-M |
Synonym | sodium trichloroacetate, tca-sodium, trichloroacetic acid sodium salt, antiperz, antyperz, tca sodium, acp grass killer, sodium tca, varitox, stca |
PubChem CID | 23681045 |
ChEBI | CHEBI:81976 |
IUPAC Name | sodium;2,2,2-trichloroacetate |
SMILES | C(=O)(C(Cl)(Cl)Cl)[O-].[Na+] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1700430
|
Thermo Scientific Chemicals
A1700430 |
250 g |
Each for $70.85
|
|
||||||
AAA170040B
|
Thermo Scientific Chemicals
A170040B |
1000 g |
Each for $178.42
|
|
||||||
AAA170040I
|
Thermo Scientific Chemicals
A170040I |
5000 g |
Each for $672.03
|
|
||||||
Description
Sodium trichloroacetate, is used in acid binding reagent in the textile industry in the dying process. Sodium trichloroacetate has been reported previously to be an effective denaturation reagent for proteins was applied to poly(l-lysine) and poly(l-glutamic acid) to see its effects on a coil-to-helix transition and on the chemical reactivities of the ε-amino group of poly(l-lysine). Addition of sodium trichloroacetate to poly-(l-lysine) induced a helical conformation even at neutral pH where the ε-amino group of the polymer was protonated.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium trichloroacetate, is used in acid binding reagent in the textile industry in the dying process. Sodium trichloroacetate has been reported previously to be an effective denaturation reagent for proteins was applied to poly(l-lysine) and poly(l-glutamic acid) to see its effects on a coil-to-helix transition and on the chemical reactivities of the ε-amino group of poly(l-lysine). Addition of sodium trichloroacetate to poly-(l-lysine) induced a helical conformation even at neutral pH where the ε-amino group of the polymer was protonated.
Solubility
Soluble in water, ethanol, methanol, acetone.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Hygroscopic.
Chemical Identifiers
650-51-1 | |
185.36 | |
SAQSTQBVENFSKT-UHFFFAOYSA-M | |
23681045 | |
sodium;2,2,2-trichloroacetate |
C2Cl3NaO2 | |
MFCD00064198 | |
sodium trichloroacetate, tca-sodium, trichloroacetic acid sodium salt, antiperz, antyperz, tca sodium, acp grass killer, sodium tca, varitox, stca | |
CHEBI:81976 | |
C(=O)(C(Cl)(Cl)Cl)[O-].[Na+] |
Specifications
650-51-1 | |
C2Cl3NaO2 | |
250 g | |
3572664 | |
14,9627 | |
Soluble in water,ethanol,methanol,acetone. | |
C(=O)(C(Cl)(Cl)Cl)[O-].[Na+] | |
185.36 | |
CHEBI:81976 | |
97% |
>300°C | |
MFCD00064198 | |
UN3077 | |
Hygroscopic | |
sodium trichloroacetate, tca-sodium, trichloroacetic acid sodium salt, antiperz, antyperz, tca sodium, acp grass killer, sodium tca, varitox, stca | |
SAQSTQBVENFSKT-UHFFFAOYSA-M | |
sodium;2,2,2-trichloroacetate | |
23681045 | |
185.37 | |
Sodium trichloroacetate |
Safety and Handling
GHS H Statement
H335
May cause respiratory irritation.
P261-P271-P304+P340-P312-P501c
H335
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 211-479-2
RTECSNumber : AJ9100000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only