missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium trimetaphosphate, Thermo Scientific Chemicals

$124.01 - $566.11

Chemical Identifiers

CAS 7785-84-4
Molecular Formula Na3O9P3
Molecular Weight (g/mol) 305.882
MDL Number MFCD00867826
InChI Key UGTZMIPZNRIWHX-UHFFFAOYSA-K
Synonym sodium trimetaphosphate, trisodium trimetaphosphate, trisodium metaphosphate, cyclic sodium trimetaphosphate, unii-3ih6169rl0, sodium phosphate napo3 3, sodium trimetaphosphate usan, metaphosphoric acid, trisodium salt, sodium metaphosphate na3 p3o9, trisodium trimetaphosphate na3p3o9
PubChem CID 24579
IUPAC Name trisodium;2,4,6-trioxido-1,3,5,2$l^{5},4$l^{5},6$l^{5}-trioxatriphosphinane 2,4,6-trioxide
SMILES [O-]P1(=O)OP(=O)(OP(=O)(O1)[O-])[O-].[Na+].[Na+].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA89063A1
View Documents
Thermo Scientific Chemicals
089063A1
1 kg
Each for $124.01
 
AA89063A7
View Documents
Thermo Scientific Chemicals
089063A7
5 kg
Each for $566.11
 
Description

Description

Sodium trimetaphosphate is used as a crosslinking agent in the cross-linked amylase-resistant starch. It acts as an intermediate in food industries. Further, it serves as a corrosion inhibitors, anti-scaling agents, fillers, finishing agents, plating agents and surface treating agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7785-84-4
305.882
UGTZMIPZNRIWHX-UHFFFAOYSA-K
24579
[O-]P1(=O)OP(=O)(OP(=O)(O1)[O-])[O-].[Na+].[Na+].[Na+]
Na3O9P3
MFCD00867826
sodium trimetaphosphate, trisodium trimetaphosphate, trisodium metaphosphate, cyclic sodium trimetaphosphate, unii-3ih6169rl0, sodium phosphate napo3 3, sodium trimetaphosphate usan, metaphosphoric acid, trisodium salt, sodium metaphosphate na3 p3o9, trisodium trimetaphosphate na3p3o9
trisodium;2,4,6-trioxido-1,3,5,2$l^{5},4$l^{5},6$l^{5}-trioxatriphosphinane 2,4,6-trioxide

Specifications

7785-84-4
1 kg
MFCD00867826
Soluble in water. Insoluble in alcohol.
[O-]P1(=O)OP(=O)(OP(=O)(O1)[O-])[O-].[Na+].[Na+].[Na+]
305.882
305.89
Sodium trimetaphosphate
Powder
Na3O9P3
sodium trimetaphosphate, trisodium trimetaphosphate, trisodium metaphosphate, cyclic sodium trimetaphosphate, unii-3ih6169rl0, sodium phosphate napo3 3, sodium trimetaphosphate usan, metaphosphoric acid, trisodium salt, sodium metaphosphate na3 p3o9, trisodium trimetaphosphate na3p3o9
UGTZMIPZNRIWHX-UHFFFAOYSA-K
trisodium;2,4,6-trioxido-1,3,5,2$l^{5},4$l^{5},6$l^{5}-trioxatriphosphinane 2,4,6-trioxide
24579
2.49
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 232-088-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only