Learn More
Sodium vinylsulfonate, tech. ca 25% aq. soln.
CAS: 3039-83-6 | C2H3NaO3S | 130.09 g/mol
$66.73 - $361.89
Chemical Identifiers
| CAS | 3039-83-6 |
|---|---|
| Molecular Formula | C2H3NaO3S |
| Molecular Weight (g/mol) | 130.09 |
| MDL Number | MFCD00007520 |
| InChI Key | BWYYYTVSBPRQCN-UHFFFAOYSA-M |
| Synonym | sodium ethenesulfonate, sodium vinylsulfonate, ethenesulfonic acid, sodium salt, sodium ethylenesulfonate, sodium vinyl sulfonate, sodium ethylenesulphonate, sodium apolate, unii-f7k3l38z7b, sodium ethenesulfonate in h2o, lyapolate sodium usan |
| PubChem CID | 3270262 |
| IUPAC Name | sodium;ethenesulfonate |
| SMILES | [Na+].[O-]S(=O)(=O)C=C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL15356AE
|
Thermo Scientific Chemicals
L15356AE |
100 mL |
Each for $66.73
|
|
|||||
|
AAL15356AP
|
Thermo Scientific Chemicals
L15356AP |
500 mL |
Each for $229.49
|
|
|||||
|
AAL15356AU
|
Thermo Scientific Chemicals
L15356AU |
1000 mL |
Each for $361.89
|
|
|||||
Description
Sodium vinylsulfonate is a useful reagent (monomer) for the formation of poly(anionic) polymers and copolymers. It is employed as a basic brightener and leveling agent in nickel baths. It is also used as intermediate for organic synthesis, surfactant, pharmaceutical industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium vinylsulfonate is a useful reagent (monomer) for the formation of poly(anionic) polymers and copolymers. It is employed as a basic brightener and leveling agent in nickel baths. It is also used as intermediate for organic synthesis, surfactant, pharmaceutical industry.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to light. Incompatible with oxidizing agents.
Chemical Identifiers
| 3039-83-6 | |
| 130.09 | |
| BWYYYTVSBPRQCN-UHFFFAOYSA-M | |
| 3270262 | |
| [Na+].[O-]S(=O)(=O)C=C |
| C2H3NaO3S | |
| MFCD00007520 | |
| sodium ethenesulfonate, sodium vinylsulfonate, ethenesulfonic acid, sodium salt, sodium ethylenesulfonate, sodium vinyl sulfonate, sodium ethylenesulphonate, sodium apolate, unii-f7k3l38z7b, sodium ethenesulfonate in h2o, lyapolate sodium usan | |
| sodium;ethenesulfonate |
Specifications
| 3039-83-6 | |
| C2H3NaO3S | |
| MFCD00007520 | |
| Light sensitive | |
| Soluble in water. | |
| [Na+].[O-]S(=O)(=O)C=C | |
| 130.09 | |
| 130.1 | |
| ≈25% aq. soln. |
| 1.18 | |
| 1.377 | |
| 100 mL | |
| sodium ethenesulfonate, sodium vinylsulfonate, ethenesulfonic acid, sodium salt, sodium ethylenesulfonate, sodium vinyl sulfonate, sodium ethylenesulphonate, sodium apolate, unii-f7k3l38z7b, sodium ethenesulfonate in h2o, lyapolate sodium usan | |
| BWYYYTVSBPRQCN-UHFFFAOYSA-M | |
| sodium;ethenesulfonate | |
| 3270262 | |
| Technical | |
| Sodium vinylsulfonate |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Warning
EINECSNumber : 221-242-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only