LaMotte™ Soil Macronutrients Test Kit
Make detailed evaluations of soil composition.
Supplier:  LaMotte™ LAM5928
 View more versions of this product
Catalog No. S17704
Explore available promotions
Includes:
  • Soil sampling tube
  • 50 soil sample bags
  • LaMotte Soil Handbook
  • Downloadable free resource: A Study of Soil Science
  • Soil analysis report forms
Description
Description
  • Soil analysis outfit includes all of the materials needed for accurate and detailed soil evaluation
  • Determines soil pH, nitrogen, phosphorus, potassium content and soil texture
  • Each self-contained, individual module contains the necessary materials and equipment to conduct 50 tests
  • All components are contained in a specially designed, rugged carrying case
  • Includes the LaMotte Soil Handbook and downloadable free resource, A Study of Soil Science
Octa Slide Comparator 
Test Factor: pH (40 tests)  
Range: 3.8-9.6  
Color Chart 
Test Factor: Nitrogen / Phosphorus (50 tests ea.) 
Range:  10-150 lb/acre / 10-200 lb/acre
Turbidity Column
Test Factor: Potassium (30 tests)
Range: 100-400 lb./acre
Order Info
Grades 6+.
Specifications
Specifications
Soil Macronutrients Test Kit
6
Soil Testing Station