LaMotte™ Soil Macronutrients Test Kit
Make detailed evaluations of soil composition.
Supplier:  LaMotte™ LAMR5928
 View more versions of this product
Catalog No. S17704A
Explore available promotions
Includes: Refill kit
Description
Description
  • Soil analysis outfit includes all of the materials needed for accurate and detailed soil evaluation
  • Determines soil pH, nitrogen, phosphorus, potassium content and soil texture
  • Each self-contained, individual module contains the necessary materials and equipment to conduct 50 tests
  • All components are contained in a specially designed, rugged carrying case
Order Info
Grades 6+.
Specifications
Specifications
Soil Macronutrients Test Kit
Soil Testing Station refill kit