Learn More
Solid Hard Maple Meter Sticks
Premium quality, solid North American Maple.
$23.31 - $28.67
Description
- Lacquered finish for optimum durability
- Won't dent or warp
- Scaled in millimeters on one side and inches in 0.125 in. increments on the other
- Dimensions: 1.125W × 0.31 in. thick (2.85 × 0.8cm)
Specifications
| Meter Stick | |
| Maple |