Fisherbrand™ Solid Neoprene Rubber Stoppers
Recommended where rubber would be attacked or deteriorated
 View more versions of this product
Catalog No. 14-141J
Explore available promotions
Description
Description
Recommended where rubber would be attacked or deteriorated
  • Solid neoprene
  • Resists petroleum products, fats, oils and organic chemicals
The approximate per-pack quantity shown is based on the average number of stoppers in 1 lb. (454g).
Specifications
Specifications
Green
7
7
3.7 cm
2.5 cm
Solid Stopper
Resists petroleum products, fats, oils and organic chemicals
3 cm
Neoprene