missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Synthware™ Solid Phase Peptide Synthesis Vessel, Ground Glass Joint, Side Arm
Solid Phase Peptide Synthesis Vessel with fritted glass disc and a lower ground glass joint and side connection.
$321.09 - $414.21
Specifications
For Use With (Application) | Organic Synthesis |
---|---|
Material | Borosilicate Glass |
Product Type | Vessel with T-bore PTFE stopcock and ground joint |
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
31501579
|
Synthware™
P140010M |
10 mL |
Each for $321.09
|
|
|||||
31501580
|
Synthware™
P140025M |
25 mL |
Each for $351.00
|
|
|||||
31501581
|
Synthware™
P140050M |
Each for $381.21
|
|
||||||
31501582
|
Synthware™
P140100M |
Each for $414.21
|
|
||||||
Specifications
Organic Synthesis | |
Borosilicate Glass | |
Vessel with T-bore PTFE stopcock and ground joint |