Synthware™ Solid Phase Peptide Synthesis Vessel, Ground Glass Joint, T-bore PTFE Plug
Solid Phase Peptide Synthesis Vessel with fritted glass disc and a lower ground glass joint.
Supplier:  Synthware™ P150050C
 View more versions of this product
Catalog No. 31501590
Explore available promotions
Description
Description
Specifications
Specifications
Organic Synthesis
Porosity: Coarse
Borosilicate Glass
50 mL