missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Solketal, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 153650025

 View more versions of this product

Catalog No. AC153650025


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Solketal
100-79-8
Colorless
188.0°C to 189.0°C
Authentic
Glass bottle
1.4330 to 1.4350
2.5 kg
1.06
solketal, 2,2-dimethyl-1,3-dioxolane-4-methanol, 2,2-dimethyl-1,3-dioxolan-4-yl methanol, glycerolacetone, dioxolan, isopropylidene glycerol, 1,2-isopropylideneglycerol, 1,2-o-isopropylideneglycerol, 1,3-dioxolane-4-methanol, 2,2-dimethyl, glycerol dimethylketal
RNVYQYLELCKWAN-UHFFFAOYSA-N
(2,2-dimethyl-1,3-dioxolan-4-yl)methanol
7528
132.16
Liquid
97%
-50°C
1.0600g/mL
90°C
96% min. (GC)
C6H12O3
MFCD00063238
19, V,2, 362
15, 5259
Solubility in water: miscible. Other solubilities: miscible with alcohol, acetals, esters, ethers,, aromatic hydrocarbons, chlorinated hydrocarbons,, gasolines and petr.ether
CC1(OCC(O1)CO)C
132.16
11 mPa.s (20°C)
97%

Chemical Identifiers

100-79-8
132.16
RNVYQYLELCKWAN-UHFFFAOYSA-N
7528
CC1(OCC(O1)CO)C
C6H12O3
MFCD00063238
solketal, 2,2-dimethyl-1,3-dioxolane-4-methanol, 2,2-dimethyl-1,3-dioxolan-4-yl methanol, glycerolacetone, dioxolan, isopropylidene glycerol, 1,2-isopropylideneglycerol, 1,2-o-isopropylideneglycerol, 1,3-dioxolane-4-methanol, 2,2-dimethyl, glycerol dimethylketal
(2,2-dimethyl-1,3-dioxolan-4-yl)methanol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.

GHS P Statement
Wear eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
If eye irritation persists: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 202-888-7

RTECSNumber : JI0400000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only