missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Solketal, 97%, Thermo Scientific Chemicals
$70.13 - $547.27
Chemical Identifiers
CAS | 100-79-8 |
---|---|
Molecular Formula | C6H12O3 |
Molecular Weight (g/mol) | 132.16 |
MDL Number | MFCD00063238 |
InChI Key | RNVYQYLELCKWAN-UHFFFAOYSA-N |
Synonym | solketal, 2,2-dimethyl-1,3-dioxolane-4-methanol, 2,2-dimethyl-1,3-dioxolan-4-yl methanol, glycerolacetone, dioxolan, isopropylidene glycerol, 1,2-isopropylideneglycerol, 1,2-o-isopropylideneglycerol, 1,3-dioxolane-4-methanol, 2,2-dimethyl, glycerol dimethylketal |
PubChem CID | 7528 |
IUPAC Name | (2,2-dimethyl-1,3-dioxolan-4-yl)methanol |
SMILES | CC1(OCC(O1)CO)C |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC153651000
|
Thermo Scientific Chemicals
153651000 |
100 g | Glass bottle |
Each for $70.13
|
|
||||
AC153650025
|
Thermo Scientific Chemicals
153650025 |
2.5 kg | Glass bottle |
Each for $547.27
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
100-79-8 | |
132.16 | |
RNVYQYLELCKWAN-UHFFFAOYSA-N | |
7528 | |
CC1(OCC(O1)CO)C |
C6H12O3 | |
MFCD00063238 | |
solketal, 2,2-dimethyl-1,3-dioxolane-4-methanol, 2,2-dimethyl-1,3-dioxolan-4-yl methanol, glycerolacetone, dioxolan, isopropylidene glycerol, 1,2-isopropylideneglycerol, 1,2-o-isopropylideneglycerol, 1,3-dioxolane-4-methanol, 2,2-dimethyl, glycerol dimethylketal | |
(2,2-dimethyl-1,3-dioxolan-4-yl)methanol |
Specifications
100-79-8 | |
Colorless | |
188.0°C to 189.0°C | |
Authentic | |
Glass bottle | |
1.4330 to 1.4350 | |
100 g | |
1.06 | |
solketal, 2,2-dimethyl-1,3-dioxolane-4-methanol, 2,2-dimethyl-1,3-dioxolan-4-yl methanol, glycerolacetone, dioxolan, isopropylidene glycerol, 1,2-isopropylideneglycerol, 1,2-o-isopropylideneglycerol, 1,3-dioxolane-4-methanol, 2,2-dimethyl, glycerol dimethylketal | |
RNVYQYLELCKWAN-UHFFFAOYSA-N | |
(2,2-dimethyl-1,3-dioxolan-4-yl)methanol | |
7528 | |
132.16 | |
Liquid |
-50°C | |
1.0600g/mL | |
90°C | |
96% min. (GC) | |
C6H12O3 | |
MFCD00063238 | |
19, V,2, 362 | |
15, 5259 | |
Solubility in water: miscible. Other solubilities: miscible with alcohol, acetals, esters, ethers,, aromatic hydrocarbons, chlorinated hydrocarbons,, gasolines and petr.ether | |
CC1(OCC(O1)CO)C | |
132.16 | |
11 mPa.s (20°C) | |
97% | |
Solketal, 97% |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS P Statement
Wear eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
If eye irritation persists: Get medical advice/attention.
GHS Signal Word: Warning
EINECSNumber : 202-888-7
RTECSNumber : JI0400000
TSCA : TSCA
RUO – Research Use Only