missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
PYREX™ Solvent Still Head Distilling Apparatus, Standard Taper Joints with 2mm T-bore PTFE stopcock
Apparatus for maintaining oxygen-free solvents
Supplier: PYREX™ 3575250
Description
Apparatus for maintaining oxygen free solvents. Head has sufficient storage space to allow solvent removal by syringe or drainage through 2 mm T-bore PTFE stopcock. With standard taper 24/40 outer joint at top, standard taper 24/40 inner joint at bottom and standard taper 14/20 inner joint on drainage arm.
Specifications
245 mm | |
Solvent Still Head Distilling Apparatus | |
9.64 in. | |
250 mL | |
8.4 oz. |
For maintaining oxygen free solvents | |
Pyrex™ | |
8.4 oz., 250 mL | |
2 |