Kent Scientific SomnoSuite™ Module Upgrade: MouseSTAT™ Pulse Oximeter and Heart Rate Monitor
For use with SomnoSuite Low-Flow Anesthesia System with Integrated Digital Vaporizer.
Supplier: Kent Scientific SS03MODULE
Includes: Mouse sensor
Description
- For use on mice and rats
- Soft touch paw sensors reduces pressure to maintain unrestricted blood flow
- Modular capability, integrates with vaporizer, ventilation, and end-tidal CO2 monitoring modules
Specifications
- Sensors: Mouse, rat, and large animal
- Heart Rate: Up to 900bpm
Applications
Vital sign monitoring, pulse oximetry, heart rate, physiological monitoring, anesthesia, surgery
Specifications
Mouse, Rat | |
Pulse Oximeter and Heart Rate Monitor Module Upgrade | |
Soft touch paw sensor for rat, mouse and large animal |
Vital Sign Monitoring, Pulse Oximetry, Heart Rate, Physiological Monitoring, Anesthesia, Surgery | |
SomnoSuite Low-Flow Anesthesia System with Integrated Digital Vaporizer |