missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytiva Sonicated DNAs for Hybridization Studies
Suitable for blocking non-specific binding sites in hybridization experiments to reduce background. Sonicated DNAs for Hybridization Studies are purified by phenol extraction and freeze dried.
$716.00 - $865.00
Specifications
Format | Solid |
---|---|
Content And Storage | -20°C |
Quantity | 100/Pk. |
For Use With (Application) | Hybridisation experiments |
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
45-001-362
|
Cytiva
27456301 |
Sonicated Calf Thymus DNA |
Each for $865.00
|
|
|||||
45-001-363
|
Cytiva
27456501 |
Sonicated Salmon Sperm DNA |
Each for $716.00
|
|
|||||
Description
- Fragmented DNA with a size range from 200-6000 nucleotides
- Purified by phenol extraction and freeze dried
- Tested for absence of nuclease
- DNA isolated from Calf thymus is sonicated to fragment the DNA to an average size of approximately 3000 bp
Specifications
Solid | |
-20°C | |
100/Pk. | |
Hybridisation experiments |