Branson Ultrasonics™ Sonifier™ SFX150 Cell Disruptor
Branson Ultrasonics Sonifier™ SFX150 Cell Disruptor with advanced controls gives more precise control over all critical elements of experiment.
Supplier: Branson Ultrasonics™ 1010631096R
Description
Easy-to-use interface allows in-process monitoring of key variables, including time, power, energy, temperature (requires optional temperature probe) and experiment progress.- Processing up to 150mL
- Quiet to operate without a sound enclosure
- True temperature control (requires separate temp probe)
- Continuous or pulsed ultrasonics
- Time or energy mode with energy pulsing
- Experiment progress indicator
Specifications
Cell Disruptor with Handheld Converter | |
150 W | |
Sample preparation, cell disruption, nanoparticle disperson, mixing, homogenization, DNA, cell lysis, protein extration | |
SFX150 | |
6.3 kg |
120 V | |
31.8 x 19.1 x 23.5 cm (12.5 x 7.5 x 9.25 in.) | |
Power supply, 40 kHz converter, 3.17 mm (0.125 in.) microtip, power cord, manual, wrench set | |
14 lb. |