Sonoco™ ThermoSafe DiagnosticShipper Systems, Without Tubes
Ready-to-use combination/triple packaging kits
$469.70 - $713.82
Specifications
Certifications/Compliance | Conforms with DOT, USPS, IATA packing instruction 650 |
---|---|
Applications | Diagnostic specimen shipping system without tubes |
Catalog Number | Mfr. No. | Includes | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity | |||||
03-52-556
|
Sonoco™
570 |
Plastic envelope, absorbent material, bubble wrap |
Pack of 25 Each for $614.38
|
|
|||||
03-52-557
|
Sonoco™
571 |
Two plastic envelopes, absorbent materials, bubble wraps |
Pack of 15 Each for $713.82
|
|
|||||
03-528-25
|
Sonoco™
474 |
Absorbent material, tape, poly bag, carton |
Pack of 33 Each for $469.70
|
|
|||||
Description
Tested and certified to meet the material and performance requirements for the transport of diagnostic specimens in conformance with U.S. Department of Transportation (DOT), U.S. Postal Service and International Air Transport Association (IATA) Packing Instruction 650.
Specifications
Conforms with DOT, USPS, IATA packing instruction 650 | |
Diagnostic specimen shipping system without tubes |