Sonoco™ ThermoSafe Envoy™ Global Pre-Qualified Insulated Shippers with Winter Packout
Temperature controlled packaging solution
Supplier:  Sonoco™ E18C2W
 View more versions of this product
Catalog No. 03-531-713
Explore available promotions
Description
Description
  • Winter packout configuration
  • Positioned as low material packaging cost solution
  • Offers various sizes, temperature ranges and flexibility
  • Envoy Global line is able to change from controlled room temperature, refrigerated and frozen temperatures by changing the refrigerants
  • Simple and error-free
  • Utilizes four EPS container sizes
  • Pre-Qualified to ISCsilver ambient profile
  • Zero Bench Time refrigerant design
  • Expanded polystyrene insulation
  • Low packaging material cost
  • Simple designs
  • Air flow channels built into EPS, allows for less refrigerant needed in each packout
Duration
  • Refrigerated and CRT: Minimum of two days
  • Frozen: Minimum of four days
Shipping pharmaceuticals, clinical treatments and diagnostic samples
Specifications
Specifications
8 Pallets
521 x 483 x 527 mm (20.5 x 19 x 20.7 in.)
19.4 lb.
18.2 L
Expanded Polystyrene
Insulated Shipper with Winter Packout Configuration
4.8 gal.
8.8 kg
CRT: 15° to 25°C