Sonoco™ ThermoSafe Infectious Shipper
Ideal for transporting infectious substances and meet U.S. Department of Transportation (DOT) and IATA requirements for transporting infectious substances.
$654.58 - $654.58
Specifications
Includes Compartmentalized bubble pouch
Certifications/Compliance DOT Regulation 49 CFR Parts 171 to 178 and IATA Packing Instructions 602
View More Specs
Products 1
Catalog Number Mfr. No. Certifications/Compliance Dimensions (LxWxH) Price Quantity  
Catalog Number Mfr. No. Certifications/Compliance Dimensions (LxWxH) Price Quantity  
03-600-148
View Documents
Sonoco™
INF2000
DOT Regulation 49 CFR Parts 171 to 178 and IATA Packing Instructions 602 4 x 4 x 6.75 in. (10 x 10 x 17 cm)
Pack of 6 Each for $654.58
Description
Description
Consists of:
  • Foam container molded of high-density expanded polystyrene
  • Absorbent pad
  • Screw cap aluminum can measuring 3D × 6 in.H (7.6 × 15cm)
  • Flexible foam insert with four pre-drilled holes to accommodate supplied test tubes
  • Four 10mL sterile blood collection tubes (16 × 100mm)
  • Press-lock, clear plastic bag
  • Corrugated carton
  • Packaging instructions
Compliance
Meets the specifications for testing and performance by D.O.T. Regulation 49 CFR Parts 178 to 179.
Specifications
Specifications
Compartmentalized bubble pouch
DOT Regulation 49 CFR Parts 171 to 178 and IATA Packing Instructions 602
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'