Sonoco™ ThermoSafe InsulatedShipper-PUR, Packed in Pallet
High performance polyurethane containers meet the needs of the most demanding temperature sensitive payloads
$224.85 - $4498.00
Specifications
For Use With (Application) Protects highly temperature sensitive shipments from moderate and extreme ambient temperature exposure
View More Specs
Products 5
Catalog Number Mfr. No. Dimensions (L x W x H) Price Quantity  
Catalog Number Mfr. No. Dimensions (L x W x H) Price Quantity  
03530052
View Documents Promotion Details
Sonoco™
E89
21 x 14 x 17.5 in. (53.3 x 35.5 x 44.4 cm)
Each for $224.85
Case of 18 Each for $4,047.30
03-532-112
View Documents Promotion Details
Sonoco™
E438
29 x 24.25 x 24.25 in. (73.6 x 61.5 x 61.5 cm)
Case of 4 Each for $3,875.27
03-532-129
View Documents Promotion Details
Sonoco™
TSE126
19 x 14.75 x 22.25 in. (48.2 x 37.4 x 56.5 cm)
Case of 18 Each for $4,498.00
03-532-133
View Documents Promotion Details
Sonoco™
E174
20.25 x 20.25 x 24.5 in. (51.4 x 51.4 x 62.2 cm)
Case of 8 Each for $3,249.48
03-530-205
View Documents Promotion Details
Sonoco™
E327
23.25 x 23 x 24 in. (59 x 58.42 x 60.9 cm)
Case of 8 Each for $2,596.48
Description
Description
Polyurethane insulation offers greater insulating power than equivalent thicknesses of EPS containers. Thick walls and 200 lb. outer corrugate offer protective shock absorption and cushioning during shipping and handling.
  • Protects highly temperature sensitive shipments from moderate and extreme ambient temperature exposure
  • Provides extensive protection for long range journeys
  • Durable one-piece closed-cell polyurethane foam insulated shippers are CFC and HCFC-free—comply with international clean air regulations
  • Proprietary process laminates highly efficient insulation core to inner and outer corrugate walls
  • Withstands crush compression in excess of 5000 lb.
  • Flexible polyurethane foam plug seals the top, cushions and insulates contents, vents dry ice
  • R-value is +1.5 times EPS R-value, per inch of thickness
Specifications
Specifications
Protects highly temperature sensitive shipments from moderate and extreme ambient temperature exposure
SDS
missing translation for 'documents'
missing translation for 'documents'