missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sorbic acid, free acid, ≥99%, MP Biomedicals™
Supplier: MP Biomedicals Inc 0210293780
Specifications
Sorbic acid, free acid | |
Yellow | |
C6H8O2 | |
100 g | |
WSWCOQWTEOXDQX-MQQKCMAXSA-N | |
(2E,4E)-hexa-2,4-dienoic acid | |
643460 | |
112.13 | |
Crystalline Powder |
110-44-1 | |
1.2g/cm3 at 20°C | |
MFCD00002703 | |
sorbic acid, 2,4-hexadienoic acid, 2e,4e-hexa-2,4-dienoic acid, 2e,4e-hexadienoic acid, panosorb, 2-propenylacrylic acid, trans,trans-sorbic acid, sorbistat, hexadienoic acid, e,e-2,4-hexadienoic acid | |
C\C=C\C=C\C(O)=O | |
112.13 | |
CHEBI:38358 | |
≥99% |
Chemical Identifiers
110-44-1 | |
112.13 | |
WSWCOQWTEOXDQX-MQQKCMAXSA-N | |
643460 | |
(2E,4E)-hexa-2,4-dienoic acid |
C6H8O2 | |
MFCD00002703 | |
sorbic acid, 2,4-hexadienoic acid, 2e,4e-hexa-2,4-dienoic acid, 2e,4e-hexadienoic acid, panosorb, 2-propenylacrylic acid, trans,trans-sorbic acid, sorbistat, hexadienoic acid, e,e-2,4-hexadienoic acid | |
CHEBI:38358 | |
C\C=C\C=C\C(O)=O |