missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sotalol hydrochloride, 98%, Thermo Scientific Chemicals

Potent beta adrenergic antagonist

$130.03 - $1566.86

Chemical Identifiers

CAS 959-24-0
Molecular Formula C12H21ClN2O3S
Molecular Weight (g/mol) 308.82
MDL Number MFCD00242937
InChI Key VIDRYROWYFWGSY-UHFFFAOYNA-N
Synonym sotalol hydrochloride, sotalol hcl, betapace, betapace af, sotacor, sotalex, berlex, mead johnson 1999, sorine
PubChem CID 66245
ChEBI CHEBI:9207
IUPAC Name hydrogen N-(4-{1-hydroxy-2-[(propan-2-yl)amino]ethyl}phenyl)methanesulfonamide chloride
SMILES [H+].[Cl-].CC(C)NCC(O)C1=CC=C(NS(C)(=O)=O)C=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ63772MC
View Documents
Thermo Scientific Chemicals
J63772MC
100 mg
Each for $130.03
 
AAJ6377203
View Documents
Thermo Scientific Chemicals
J6377203
1 g
Each for $949.62
 
AAJ6377206
View Documents
Thermo Scientific Chemicals
J6377206
5 g
Each for $1,566.86
 
Description

Description

Sotalol hydrochloride is used as a potent beta adrenergic antagonist, prolongs the action potential and increases the refractory period. Sotalol hydrochloride is also considered a non-selective β blocker and a potassium channel blocker with an IC50 of 43 μM.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sotalol hydrochloride is used as a potent beta adrenergic antagonist, prolongs the action potential and increases the refractory period. Sotalol hydrochloride is also considered a non-selective β blocker and a potassium channel blocker with an IC50 of 43 µM.

Solubility
Soluble in phosphate buffered saline, DMSO, ethanol, water, and methanol.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

959-24-0
308.82
VIDRYROWYFWGSY-UHFFFAOYNA-N
66245
hydrogen N-(4-{1-hydroxy-2-[(propan-2-yl)amino]ethyl}phenyl)methanesulfonamide chloride
C12H21ClN2O3S
MFCD00242937
sotalol hydrochloride, sotalol hcl, betapace, betapace af, sotacor, sotalex, berlex, mead johnson 1999, sorine
CHEBI:9207
[H+].[Cl-].CC(C)NCC(O)C1=CC=C(NS(C)(=O)=O)C=C1

Specifications

959-24-0
C12H21ClN2O3S
100 mg
sotalol hydrochloride, sotalol hcl, betapace, betapace af, sotacor, sotalex, berlex, mead johnson 1999, sorine
VIDRYROWYFWGSY-UHFFFAOYNA-N
hydrogen N-(4-{1-hydroxy-2-[(propan-2-yl)amino]ethyl}phenyl)methanesulfonamide chloride
66245
308.82
Crystalline Powder
White
MFCD00242937
14,8728
Soluble in phosphate buffered saline,DMSO,ethanol,water,and methanol.
[H+].[Cl-].CC(C)NCC(O)C1=CC=C(NS(C)(=O)=O)C=C1
308.82
CHEBI:9207
98%
Sotalol hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 213-496-0

RTECSNumber : PB0826000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only