missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Eisco™ Soxhlet Extraction Apparatus

Sets consist of Soxhlet extractor bulb, condenser, and flask, each with Interchangeable standard ground joints

Supplier:  Eisco™ CH0888C

 View more versions of this product

Catalog No. S29511


Explore available promotions

Description

Description

  • Soxhlet extraction apparatus
  • Made from high quality borosilicate 3.3 glass
  • Feature interchangeable standard ground joint
  • Each set includes: 1 Soxhlet extractor bulb, 1 condenser, and 1 flask
  • Specifications

    Specifications

    1
    Borosilicate Glass
    Soxhlet extractor bulb, condenser, flask
    12 cm
    200 mL
    Distillation
    50.1 cm
    Videos
    SDS
    Documents

    Documents

    Promotions

    Promotions

    1 year limited manufacturer warranty