Learn More
SP H-B Instrument™ Accredited Thermometer Calibration Services: ISO 17025:2005
Meets the requirements of ISO, ANSI and NIST standards. Thermometer Calibration Services can be used to calibrate new thermometers as well as to recalibrate previously calibrated instruments. Report includes correction values. All laboratory thermometer brands accepted.
$940.50 - $2676.11
Specifications
For Use With (Equipment) Any brand Liquid-in-Glass Thermometer
Certifications/Compliance ISO 17025, ANSI NCSL-Z-540, NIST
Description Accredited calibration/recalibration service for LIG thermometers
Includes Calibration report, data storage, 1 yr reminder
View More Specs
Includes: Thermometer calibration service; available for all brands of thermometer.
Products 13
Catalog Number Mfr. No. Calibration Type Duration Price Quantity  
Catalog Number Mfr. No. Calibration Type Duration Price Quantity  
13-202-693
View Documents
N/A Temperature: 100°C to 150°C 10 Day Turnaround (avg.)
Each for $940.50
Only null left
13-202-689
View Documents
N/A Temperature: -30°C to -1°C 10 Day Turnaround (avg.)
Each for $940.50
Only null left
13-202-694
View Documents
N/A Temperature: 100°C to 150°C 10 Day Turnaround (avg.)
Each for $1,410.14
Only null left
13-202-686
View Documents
SP H-B Instrument™
B601210000
Temperature: 1 to 100°C 5 Day Turnaround (avg.)
Each for $951.22
Only null left
13-202-690
View Documents
N/A Temperature: -30°C to -1°C 10 Day Turnaround (avg.)
Each for $1,384.21
Only null left
13-202-698
View Documents
N/A Temperature: -80°C to -31°C 10 Day Turnaround (avg.)
Each for $1,779.43
Only null left
13-202-695
View Documents
N/A Temperature: 100°C to 150°C 10 Day Turnaround (avg.)
Each for $1,571.93
Only null left
13-202-703
View Documents
SP H-B Instrument™
B601500000
Temperature: 150°C to 200°C 10 Day Turnaround (avg.)
Each for $1,493.94
Only null left
13-202-691
View Documents
N/A Temperature: -30°C to -1°C 10 Day Turnaround (avg.)
Each for $1,934.29
Only null left
13-202-699
View Documents
N/A Temperature: -80°C to -31°C 10 Day Turnaround (avg.)
Each for $1,707.36
Only null left
13-202-688
View Documents
N/A Temperature: 1 to 100°C 5 Day Turnaround (avg.)
Each for $1,656.43
Only null left
13-202-692
View Documents
N/A Temperature: -30°C to -1°C 10 Day Turnaround (avg.)
Each for $1,961.93
Only null left
13-202-700
View Documents
N/A Temperature: -80°C to -31°C 10 Day Turnaround (avg.)
Each for $2,676.11
Only null left
Description
Description
Properly calibrated instruments are required to adhere to laboratory standard operating procedures. Many factors can cause drift from accepted values. In general laboratory use, it is recommended that liquid-in-glass thermometers be calibrated at one year intervals (NIST recommends six month intervals for LIG thermometers used as standards).
  • Accredited calibration documentation meets the requirements of ISO 17025 and ANSI NCSL-Z-540
  • Calibration is performed in SP H-B Instrument™ exclusive accredited/registered ISO/IEC 17025:2005, A2LA accredited laboratory (Cert No. 2448.01)
  • Calibration is performed against equipment whose calibration is traceable to NIST
  • Average 5 day turn-around for thermometer calibration points between 0 and 100°C (32 to 212°F); points outside that range can take up to 10 working days to complete
  • The calibration report with correction values will be sent and will also be stored up to 5 years
  • A 1-year recalibration reminder will be sent to ensure reliable and accurate instrument readings throughout the life of the equipment
Instructions for Ordering/Receiving Calibration Services:
  1. Contact Fisher Scientific at 800-766-7000 to let them know the type of instrument you will be sending in, or the catalog number of the item that you would like to order, and the required calibration points. The representative will provide you with the cost and place the order.
  2. If you are sending in a piece of equipment, the representative will provide you with a Return Material Authorization (RMA) number. Securely pack and send your instrument(s) with the RMA number on the outside of the box to: H-B Instrument – SP, 1002 Harding Highway, Buena NJ 08310, Attn. Calibration.
  3. SP H-B Instrument™ Calibration Laboratory will perform the calibration and send you the calibration report along with the instrument.
Specifications
Specifications
Any brand Liquid-in-Glass Thermometer
ISO 17025, ANSI NCSL-Z-540, NIST
Accredited calibration/recalibration service for LIG thermometers
Calibration report, data storage, 1 yr reminder
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'