Bel-Art™ Acetal Plastic Spring-Style Tubing Clamps
Self-closing spring operated tubing clamp is suitable for all flexible tubing.
Supplier: Bel-Art™ H182130000EMD
Description
- Positive-action clamp opens by squeezing handles together
- Naturally clamps shut to stop flow
- Chemical- and stain-resistant
- Made of Acetal plastic and stainless steel
Specifications
3 | |
Acetal Plastic | |
White | |
8 mm |
Spring Style Tubing Clamp | |
For tubing up to 8 mm (5/16 in.) O.D. | |
Angled |