missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bel-Art™ SP Scienceware™ LabMat™ Bench Liners
Simplify cleanup and safeguard work surfaces from stains, spills and wear. Bel Art™ SP Scienceware™ LabMat™ Bench Liners can be rolled up and discard after contamination.
$150.54 - $473.68
Specifications
Product Type | Bench Protector |
---|---|
Material | Polyethylene |
Absorbancy | 400 mL/m2 |
Width (English) | 20 in. |
Width (Metric) | 508 mm |
Catalog Number | Mfr. No. | Color | Dimensions (L x W) | Description | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Dimensions (L x W) | Description | Price | Quantity | |||
11-999-19
|
Bel-Art™
F246751000 |
Orange | 50 ft. x 20 in. (15.2m x 508mm) | One single roll, 50 ft. |
Each for $150.54
Case of 6 Each for $861.54
|
|
|||
11-999-20
|
Bel-Art™
F246750020 |
White | 24 x 20 in. (610 x 508mm) | Pack of 100 sheets |
Pack of 100 Each for $473.68
|
|
|||
11-999-18
|
Bel-Art™
246750000 |
White | 50 ft. x 20 in. (15.2m x 508mm) | One single roll, 50 ft. |
Each for $473.68
Case of 6 Each for $1,164.54
|
|
|||
Description
- Highly absorbent upper surface traps up to 420mL/m2 of water, dyes, corrosive chemicals and radioactive solutions
- Flexible polyethylene film backing is nonskid and prevents soak-through of most solutions
- Cut to size easily with scissors or the Scienceware™ Safety Cutter (Mfr. No. 21012-0000)
- Roll up and discard in proper waste container after contamination
- Select orange to identify use with biohazard materials
Specifications
Bench Protector | |
400 mL/m2 | |
508 mm | |
Simplify cleanup and safeguard work surfaces from stains, spills and wear |
Polyethylene | |
20 in. | |
Single-use |