missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Polyethylene, Carboy with Spigot

For Use With: For dispense cleaning solutions, laboratory chemicals, disinfectants, and intermediate phase mixtures and slurries for various manufacturing processes; Closure Material: Polyethylene; Color: White

$401.90 - $401.90

Specifications

Color White
For Use With (Application) For dispense cleaning solutions, laboratory chemicals, disinfectants, and intermediate phase mixtures and slurries for various manufacturing processes
Product Type Carboy with Spigot
View More Specs

Products 1
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
01-189-0112
View Documents Promotion Details
Bel-Art™
F118470010
4 L
Each for $401.90
Case of 6 Each for $2,411.40
 
Description

Description

  • Lighter than glass and easy to handle
  • Heavy knurling on cap provides excellent grip for opening and closing
  • Resistant to corrosive acids and alkalis
  • The spigot requires only half turn to open or close
  • Smooth sides accept adhesive labels and can be marked with most indelible markers

Use to dispense cleaning solutions, laboratory reagents, disinfectants, and intermediate phase mixtures and slurries for various manufacturing processes.

Specifications

Specifications

White
For dispense cleaning solutions, laboratory chemicals, disinfectants, and intermediate phase mixtures and slurries for various manufacturing processes
Carboy with Spigot
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'