missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Polypropylene, Vacuum Aspirator Bottle

For Use With: For effortless, continuous vacuum aspiration of a variety of liquids, including tissue culture media, supernatants, and chemical wastes.; Autoclavable; Closure Material: Polypropylene; Color: Blue (Translucent)

$1030.10 - $1129.40

Specifications

Autoclavable Autoclavable
Closure Material Polypropylene
Color Blue (Translucent)
For Use With (Application) For effortless, continuous vacuum aspiration of a variety of liquids, including tissue culture media, supernatants, and chemical wastes.
For Use With (Equipment) For dishes, plates, flasks and tubes
View More Specs

Products 2
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
13-717-25
View Documents
Bel-Art™
F199170002
3.8 L
Each for $1,129.40
 
13-717-24
View Documents
Bel-Art™
F199170001
1.9 L
Each for $1,030.10
 
Description

Description

  • When used with a vacuum source, allows aspiration from a pipet tip, Pasteur or serological pipet inserted into the collection tube
  • Chemical and corrosion-resistant polypropylene bottle and epoxy-coated steel frame
  • Compact design does not crowd workspace or fume hood
  • Included trap prevents fluid flow into the vacuum source
  • Suitable for use with dishes, plates, flasks and tubes
  • Bottle and cap are autoclavable at 121°C (250°F)
  • Uses flexible collection tubing (1/4 in. ID x 3/8 in. OD)
Specifications

Specifications

Autoclavable
Blue (Translucent)
For dishes, plates, flasks and tubes
Vacuum Aspirator Bottle
Polypropylene
For effortless, continuous vacuum aspiration of a variety of liquids, including tissue culture media, supernatants, and chemical wastes.
Polypropylene
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'