missing translation for 'onlineSavingsMsg'
Learn More

Brady™ SPC™ MRO Plus™ Maintenance Sorbents

Highly effective for cleanup and removal of oil and water-based liquids

Supplier:  Brady™ MRO100

 View more versions of this product

Catalog No. 19-039-117


Only null left
Explore available promotions

Description

Description

  • Heavyweight material combines thickness, strength and high absorbency
  • Low-linting, so it's suitable for virtually any industrial application
  • Perforations allow for best fit
  • Incinerates to less than 0.02% ashl

Specifications

Specifications

Gray
98.4 L
Overspray leaks and drips, workstations, heavy clean-up and wipe-up , drums - liquid storage, outfalls and discharge ponds
Heavyweight
19 in.
15 in.
26 gal.
Universal Sorbent Pad
3-ply
19 x 15 in. (48.2 x 38 cm)
48.2 cm
38 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions